Läs mer
(+)-Citramalic Acid, TRC
Produktkod. 30350853
Change view
Kvantitet:
10 mg
Produktkod. Kvantitet
30350853 10 mg
1 options
Produktkod. 30350853 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC52052510MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material (+)-Citramalic Acid
CAS 6236-09-5
Anteckningar om renhetsgrad HPLC
Molekylformel C5H8O5
Kvantitet 10 mg
Synonym (2S)-2-Hydroxy-2-methylbutanedioic Acid, (+)-Methylmalic Acid, (S)-(+)-Citramalic Acid, (S)-Citramalic Acid, 2S-Methylmalic Acid
LEDER C[C@](O)(CC(=O)O)C(=O)O
Molekylvikt (g/mol) 148.11
Formel vikt 148.0372