Läs mer
Citric acid-2,4-13C2, TRC
Produktkod. 30137073
Change view
Kvantitet:
25 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30137073 100 mg
30135530 50 mg
30004018 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30137073 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC521060100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 121633-50-9
Molekylformel 13C2 C4 H8 O7
Molekylvikt (g/mol) 194.11
Synonym 2-Hydroxy-1,2,3-propane-1,3-13C2-tricarboxylic Acid
IUPAC-namn 2-hydroxy(1,3-^{13}C_{2})propane-1,2,3-tricarboxylic acid
LEDER OC(=O)[13CH2]C(O)([13CH2]C(=O)O)C(=O)O
Specifikationer
Kemiskt namn eller material Citric acid-2,4-13C2
Kvantitet 100 mg
InChI formel InChI=1S/C6H8O7/c7-3(8)1-6(13,5(11)12)2-4(9)10/h13H,1-2H2,(H,7,8)(H,9,10)(H,11,12)/i1+1,2+1
Formel vikt 194.0337