Learn More
R&D Systems™ Recombinant Human FLJ35220 GST (N-Term) Protein
Product Code. 18601731 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
10 μg
25 μg
Unit Size:
10µg
25µg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18601731 10 μg 10µg
18687530 25 μg 25µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 18601731 Supplier R&D Systems™ Supplier No. H00284131P0110UG
 Restricted Item
Beräknad leverans: 03-12-2026
A Novus product, part of R&D Systems. Recombinant Protein
This protein was produced in an in vitro wheat germ expression system that should preserve correct conformational folding that is necessary for biological function. While it is possible that this protein could display some level of activity, the functionality of this protein has not been explicitly measured or validated.
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Application) Western Blot, ELISA, Protein Array, Immunoaffinity Purification
Formulation 50 mM Tris-HCl, 10 mM reduced Glutathione, pH 8.0 in the elution buffer.
Gene ID (Entrez) 284131
Molecular Weight (g/mol) MolecularWeight-theroretical: 54.9 kDa
Quantity 10 μg
Storage Requirements Store at -80°C. Avoid freeze/thaw cycles.
Gene Alias EC 3.1, hypothetical protein FLJ35220, hypothetical protein LOC284131, MGC74455
Gene Symbol ENDOV
Product Line Novus
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Protein FLJ35220