Läs mer
Citric Acid tert-Butyl Ester, TRC
Produktkod. 30357086
Change view
Kvantitet:
25 mg
250 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30357086 250 mg
30362052 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30357086 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC521005250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 114340-52-2
Molekylformel C10H16O7
Molekylvikt (g/mol) 248.23
Synonym 3-tert-Butyl Citrate, 2-Hydroxy-1,2,3-propanetricarboxylic Acid 2-(1,1-Dimethylethyl) Ester
IUPAC-namn 3-hydroxy-3-[(2-methylpropan-2-yl)oxycarbonyl]pentanedioic acid
LEDER CC(C)(C)OC(=O)C(O)(CC(=O)O)CC(=O)O
Specifikationer
Kemiskt namn eller material Citric Acid tert-Butyl Ester
Kvantitet 250 mg
InChI formel InChI=1S/C10H16O7/c1-9(2,3)17-8(15)10(16,4-6(11)12)5-7(13)14/h16H,4-5H2,1-3H3,(H,11,12)(H,13,14)
Formel vikt 248.0896