Läs mer
(-)-Citronellal, TRC
Produktkod. 30363387
Change view
Kvantitet:
1 mL
Produktkod. Kvantitet
30363387 1 mL
1 options
Produktkod. 30363387 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC5250501ML
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material (-)-Citronellal
CAS 5949-05-3
Molekylformel C10H18O
Kvantitet 1 mL
Synonym 6-Octenal, 3,7-dimethyl-, (3S)-, 6-Octenal, 3,7-dimethyl-, (-)- (8CI), 6-Octenal, 3,7-dimethyl-, (S)-, (3S)-3,7-Dimethyl-6-octenal, (-)-(S)-Citronellal, (-)-Citronellal, (-)-β-Citronellal, (3S)-3,7-Dimethyloct-6-enal, (3S)-3,7-Dimethyloct-6-enal, (S)-(-)-3,7-Dimethyl-6-octenal, (S)-3,7-Dimethyloct-6-enal, (S)-Citronellal, L-(-)-Citronellal, L-Citronellal, S-(-)-Citronellal, (S)-(-)-Citronellal, l-Citronellal
InChI formel InChI=1S/C10H18O/c1-9(2)5-4-6-10(3)7-8-11/h5,8,10H,4,6-7H2,1-3H3/t10-/m0/s1
LEDER CC(CCC=C(C)C)CC=O
IUPAC-namn (3S)-3,7-dimethyloct-6-enal
Molekylvikt (g/mol) 154.25
Formel vikt 154.136