Läs mer
CJ-023423, TRC
Produktkod. 30007951
Change view
Kvantitet:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30007951 10 mg
30008327 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30007951 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC53580010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 415903-37-6
Molekylformel C26H29N5O3S
Molekylvikt (g/mol) 491.61
IUPAC-namn 1-[2-[4-(2-ethyl-4,6-dimethylimidazo[4,5-c]pyridin-1-yl)phenyl]ethyl]-3-(4-methylphenyl)sulfonylurea
LEDER CCc1nc2c(C)nc(C)cc2n1c3ccc(CCNC(=O)NS(=O)(=O)c4ccc(C)cc4)cc3
Specifikationer
Kemiskt namn eller material 2-Ethyl-4,6-dimethyl-1-[4-[2-[(([(4-methylphenyl)sulfonyl]amino)carbonyl)amino]ethyl]phenyl]-1H-imidazo[4,5-c]pyridine
Kvantitet 10 mg
Synonym CJ-023423, AAT 007, CJ 023423, Grapiprant, MR 10A7, N-[[[2-[4-(2-Ethyl-4,6-dimethyl-1H-imidazo[4,5-c]pyridin-1-yl)phenyl]ethyl]amino]carbonyl]-4-methylbenzenesulfonamide, RQ 00000007, RQ 7, N-[[[2-[4-(2-Ethyl-4,6-dimethyl-1H-imidazo[4,5-c]pyridin-1-yl)phenyl]ethyl]amino]carbonyl]-4-methyl-benzenesulfonamide
InChI formel InChI=1S/C26H29N5O3S/c1-5-24-29-25-19(4)28-18(3)16-23(25)31(24)21-10-8-20(9-11-21)14-15-27-26(32)30-35(33,34)22-12-6-17(2)7-13-22/h6-13,16H,5,14-15H2,1-4H3,(H2,27,30,32)