Läs mer
CK1-IN-1, MedChemExpress
Produktkod. 16497247
Encompass_Preferred
Change view
Kvantitet:
5 mg
10 mg
50 mg
Förpackningsstorlek:
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
16497247 5 mg -
16490837 50 mg 50mg
16454538 10 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 16497247 Leverantör Medchem Express Leverantörsnummer HY111820
MedChemExpress CK1-IN-1 is a casein kinase 1 (CK1) inhibitor extracted from patent WO2015119579A1, compound 1c, has IC50s of 15 nM, 16 nM, 73 nM for CK1δ, and CK1ε, p38σ MAPK, respectively.
CK1-IN-1 is a casein kinase 1 (CK1) inhibitor extracted from patent WO2015119579A1, compound 1c, has IC50s of 15 nM, 16 nM, 73 nM for CK1δ, and CK1ε, p38σ MAPK, respectively.
Specifikationer
Kemiskt namn eller material CK1-IN-1
CAS 1784751-20-7
Färg Light Yellow
Anteckningar om renhetsgrad Research
Molekylformel C24H15F2N3
Kvantitet 5 mg
Löslighetsinformation DMSO : 62.5 mg/mL (163.02 mM; Need ultrasonic) ∣H2O : < 0.1 mg/mL (insoluble)
LEDER FC1=CC=C(C2=C(C3=CC=NC=C3)NC(C4=CC=C5C=CC=CC5=C4F)=N2)C=C1
Molekylvikt (g/mol) 383.39
Formel vikt 383.39
Procent renhet 98.7%
Kvalitet Research
Fysisk form Solid
För användning med (applikation) COVID-19-anti-virus