missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ CKMT2 (Human) Recombinant Protein
Human CKMT2 full-length ORF ( AAH29140, 1 a.a. - 419 a.a.) recombinant protein with GST-tag at N-terminal.
Brand: Abnova™ H00001160-P01.25ug
Additional Details : Weight : 0.00010kg
Description
- Sequence: MASIFSKLLTGRNASLLFATMGTSVLTTGYLLNRQKVCAEVREQPRLFPPSADYPDLRKHNNCMAECLTPAIYAKLRNKVTPNGYTLDQCIQTGVDNPGHPFIKTVGMVAGDEESYEVFADLFDPVIKLRHNGYDPRVMKHTTDLDASKITQGQFDEHYVLSSRVRTGRSIRGLSLPPACTRAERREVENVAITALEGLKGDLAGRYYKLSEMTEQDQQRLIDDHFLFDKPVSPLLTCAGMARDWPDARGIWHNYDKTFLIWINEEDHTRVISMEKGGNMKRVFERFCRGLKEVERLIQERGWEFMWNERLGYILTCPSNLGTGLRAGVHVRIPKLSKDPRFSKILENLRLQKRGTGGVDTAAVADVYDISNIDRIGRSEVELVQIVIDGVNYLVDCEKKLERGQDIKVPPPLPQFGKK
Specifications
AAH29140 | |
creatine kinase, mitochondrial 2 (sarcomeric) | |
125% SDS-PAGE Stained with Coomassie Blue | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
CKMT2 | |
GST |
1160 | |
Wheat germ expression system | |
25 μg | |
SMTCK | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |