missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ CKS1B (Human) Recombinant Protein
Human CKS1B full-length ORF ( AAH07751, 1 a.a. - 79 a.a.) recombinant protein with GST-tag at N-terminal.
Brand: Abnova™ H00001163-P01.25ug
Additional Details : Weight : 0.00010kg
Description
- Sequence: MSHKQIYYSDKYDDEEFEYRHVMLPKDIAKLVPKTHLMSESEWRNLGVQQSQGWVHYMIHEPEPHILLFRRPLPKKPKK
Specifications
AAH07751 | |
CDC28 protein kinase regulatory subunit 1B | |
125% SDS-PAGE Stained with Coomassie Blue | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
CKS1B | |
GST |
1163 | |
Wheat germ expression system | |
25 μg | |
CKS1, PNAS-16, PNAS-18, ckshs1 | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |