missing translation for 'onlineSavingsMsg'
Learn More

CL-387785, MedChemExpress

Product Code. 16486537
Encompass_Preferred
Change view
Click to view available options
Kvantitet:
2 mg
5 mg
10 mg
25 mg
50 mg
100 mg
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Kvantitet
16486537 5 mg
16462866 25 mg
16424866 100 mg
16423818 10 mg
16400197 50 mg
16400086 2 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Product Code. 16486537 Supplier Medchem Express Supplier No. HY10325

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress CL-387785(EKI785; WAY-EKI 785) is an irreversible inhibitor of EGFR with IC50 of 370 pM.

CL-387785(EKI785

Specifications

Kemiskt namn eller material CL-387785
CAS 194423-06-8
Färg Light Yellow
Anteckningar om renhetsgrad Research
Molekylformel C18H13BrN4O
Kvantitet 5 mg
Synonym EKI-785 WAY-EKI 785
Löslighetsinformation DMSO : 13.67 mg/mL (35.86 mM; Need ultrasonic and warming)
LEDER O=C(NC1=CC2=C(NC3=CC=CC(Br)=C3)N=CN=C2C=C1)C#CC
Molekylvikt (g/mol) 381.23
Formel vikt 381.23
Procent renhet 98.1%
Kvalitet Research
Fysisk form Solid
För användning med (applikation) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.