Läs mer
Tocris Bioscience™ CL 82198 hydrochloride
Kvantitet:
10 mg
50 mg
Beskrivning
Selective inhibitor of MMP-13 (89% inhibition at 10 μg/mL) that displays no activity at MMP-1, MMP-9 or TACE. Inhibits in vitro invasion by the human pituitary adenoma cell line HP75. Rescues paclitaxel induced axon degradation and reduces associated neurotoxicity in zebrafish.
Specifikationer
Specifikationer
| Kemiskt namn eller material | CL 82198 hydrochloride |
| Kvantitet | 10 mg |
| Formel vikt | 338.83 |
| Renhet | >98% |
| Molekylvikt (g/mol) | 338.832 |