Läs mer
Cladinose, TRC
Produktkod. 30367168
Change view
Kvantitet:
10 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30367168 100 mg
30368744 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30367168 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC557480100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 470-12-2
Molekylformel C8H16O4
Molekylvikt (g/mol) 176.21
Synonym 2,6-Dideoxy-3-C-methyl-3-O-methyl-ribo-hexose, Azithromycin Impurity 4, (4R,5S,6S)-4-Methoxy-4,6-dimethyltetrahydro-2H-pyran-2,5-diol
IUPAC-namn (3S,4R,5R)-4,5-dihydroxy-3-methoxy-3-methylhexanal
LEDER C[C@H]([C@H]([C@](C)(CC=O)OC)O)O
Specifikationer
Kemiskt namn eller material Cladinose
Kvantitet 100 mg
InChI formel InChI=1S/C8H16O4/c1-6(10)7(11)8(2,12-3)4-5-9/h5-7,10-11H,4H2,1-3H3/t6-,7-,8+/m1/s1
Formel vikt 176.1049