Läs mer
Cladosporin, TRC
Produktkod. 30355102
Change view
Kvantitet:
0.5 mg
2.5 mg
5 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30355102 2.5 mg
30366234 5 mg
30349770 0.5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30355102 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC5575502.5MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 35818-31-6
Molekylformel C16H20O5
Molekylvikt (g/mol) 292.33
Synonym [2R-[2alpha(R*),6beta]]-3,4-dihydro-6,8-dihydroxy-3-[(tetrahydro-6-methyl-2H-pyran-2-yl)methyl]-1H-2-Benzopyran-1-one, (3R)-3,4-Dihydro-6,8-dihydroxy-3-[[(2R,6S)-tetrahydro-6-methyl-2H-pyran-2-yl]methyl]-1H-2-benzopyran-1-one, Asperentin
IUPAC-namn (3R)-6,8-dihydroxy-3-[[(2R,6S)-6-methyloxan-2-yl]methyl]-3,4-dihydroisochromen-1-one
LEDER C[C@H]1CCC[C@H](C[C@H]2Cc3cc(O)cc(O)c3C(=O)O2)O1
Specifikationer
Kemiskt namn eller material Cladosporin
Anteckningar om renhetsgrad HPLC
Kvantitet 2.5 mg
InChI formel InChI=1S/C16H20O5/c1-9-3-2-4-12(20-9)8-13-6-10-5-11(17)7-14(18)15(10)16(19)21-13/h5,7,9,12-13,17-18H,2-4,6,8H2,1H3/t9-,12+,13+/m0/s1
Formel vikt 292.1311
Procent renhet >95%