Learn More
Phenomenex™ Claricep™ Empty Flash Columns, Full Set with 2 Frits
Product Code. 31742277 Shop All Phenomenex Products
Change view
Click to view available options
Bed Weight (Metric):
3 kg
Unit Size:
Each
Product Code. Bed Weight (Metric) Unit Size
31742277 3 kg Each
1 options
Product Code. 31742277 Supplier Phenomenex™ Supplier No. FCH5000I
Beräknad leverans: 01-12-2026
Available in two flash media (irregular and spherical) with multiple chemistries, Claricep columns are optimized for resolution and high loading capacity. The deactivated silica surface promotes compound stability, excellent peak shape and high performance for both acidic and basic compounds.
  • Brand: Claricep
  • Technique: Accessory
TRUSTED_SUSTAINABILITY
Specifications
Type Empty Cartridge
Bed Weight (Metric) 3 kg