Läs mer
VITLAB™ Class A PMP Volumetric Flask
Set volumes accurately for exact measurements in lab applications with these flasks. VITLAB™ Class A PMP Volumetric Flasks feature a single calibration mark that defines the calibrated volume exactly. Available in clear or UV-absorbing, highly transparent PMP for storage of light-sensitive substances.
Brand:  VITLAB™ 67604
Produktkod. 10612882
  • ONLINE EXCLUSIVE PRICE
    324.00 SEK
    316.17 SEK / Varje
    Spara 7.83 SEK
    2% Off
Bara null kvar
Beräknad leverans: 29-12-2025
Beskrivning
  • Safely protect samples from the effects of light, while maintaining high transparency to enable volumes to be set accurately
  • With ring mark individually calibrated to ‘In’
  • Class A tolerances according to DIN EN ISO 1042
  • With printed lot number and batch certificate
  • Thermal stress up to 121°C (autoclaving) does not permanently exceed the tolerance limit
  • To preserve markings, cleaning at no higher than 60°C is recommended
  • Also available with DAkkS calibration certificate or individual quality certificate
Produktförslag
Produktförslag
Filmer
SDS
Dokument
Dokument
Certifikat
Kampanjer
Kampanjer
Korrigering av produktinnehåll
Produkttitel
VITLAB™ Class A PMP Volumetric Flask > Capacity: 1000mL; Height: 310mm; Includes: NS 24/29 stopper; Tolerance: ±0.40mL