missing translation for 'onlineSavingsMsg'
Learn More

Duran™ Class B Clear Glass Volumetric Flask

Designed for precise measurement of specified liquid amounts, preparation and storage of standard solutions.

290.00 SEK - 1305.00 SEK

Specifications

Certifications/Compliance ISO 1042, USP
Class B
Closure Material Polyethylene
Color Clear
Graduated Yes
View More Specs

Products 10
Product Code Brand Capacity (Metric) Price Quantity & Availability  
Product Code Brand Capacity (Metric) Price Quantity & Availability  
15260926 View Documents Duran™
246705458
1000 mL 880.00 SEK
Pack of 2 View alternative products
Save Up to 
»
Estimated Shipment
23-04-2020
Add to basket
 
15220926 View Documents Duran™
246702555
100 mL 371.00 SEK
Pack of 2 View alternative products
Save Up to 
»
Estimated Shipment
23-04-2020
Add to basket
 
15270926 View Documents Duran™
246706351
2000 mL 1235.00 SEK
Pack of 2 View alternative products
Save Up to 
»
Estimated Shipment
23-04-2020
Add to basket
 
15230926 View Documents Duran™
246703251
200 mL 413.00 SEK
Pack of 2 View alternative products
Save Up to 
»
Estimated Shipment
23-04-2020
Add to basket
 
15290916 View Documents Duran™
246701259
20 mL 290.00 SEK
Pack of 2 View alternative products
Save Up to 
»
Estimated Shipment
23-04-2020
Add to basket
 
15240926 View Documents Duran™
246703654
250 mL 379.00 SEK
Pack of 2 View alternative products
Save Up to 
»
Estimated Shipment
23-04-2020
Add to basket
 
15200926 View Documents Duran™
246701456
25 mL 290.00 SEK
Pack of 2 View alternative products
Save Up to 
»
Estimated Shipment
23-04-2020
Add to basket
 
15280926 View Documents Duran™
246707356
5 L 1305.00 SEK
Each View alternative products
Save Up to 
»
Estimated Shipment
23-04-2020
Add to basket
 
15250926 View Documents Duran™
246704453
500 mL 530.00 SEK
Pack of 2 View alternative products
Save Up to 
»
Estimated Shipment
23-04-2020
Add to basket
 
15210926 View Documents Duran™
246701756
50 mL 333.00 SEK
Pack of 2 View alternative products
Save Up to 
»
Estimated Shipment
23-04-2020
Add to basket
 
Description

Description

  • With one graduation mark
  • White printed image
  • Maximum recommended temperature without affecting accuracy (at 20°C): 250°C
  • Temperature resistance PE Stopper: -40° to +80°C

Calibration is based on the poured in volume (In) at a +20°C reference temperature. The volume content tolerances conform to accuracy class B, the accuracy limits of the German weights and measures regulations and DIN and ISO specifications.
Specifications

Specifications

ISO 1042, USP
Polyethylene
Yes
B
Clear
Borosilicate Glass, Duran Glass