Learn More
Abeba™ Classic 31001 Shoes
Animal fats, oil and fuel resistant. Leather/microfiber construction.
946.10 SEK
Specifications
Certifications/Compliance | CE EN ISO 20345:2011 SB A, E, FO SRC |
---|---|
Color | White |
Liner Material | Microfiber |
Material | Microfiber |
Sole Features | Slip-resistant |
Product Code | Brand | Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Size | Price | Quantity & Availability | |||||
15415714
|
Abeba™
31001/36 |
36 |
946.10 SEK
1 pair |
Estimated Shipment: 18-04-2024
Log in to see stock. |
|||||
15415724
|
Abeba™
31001/37 |
37 |
946.10 SEK
1 pair |
Estimated Shipment: 18-04-2024
Log in to see stock. |
|||||
15455724
|
Abeba™
31001/38 |
38 |
946.10 SEK
1 pair |
Estimated Shipment: 18-04-2024
Log in to see stock. |
|||||
15405734
|
Abeba™
31001/39 |
39 |
946.10 SEK
1 pair |
Estimated Shipment: 18-04-2024
Log in to see stock. |
|||||
15415734
|
Abeba™
31001/40 |
40 |
946.10 SEK
1 pair |
Estimated Shipment: 12-04-2024
Log in to see stock. |
|||||
15425734
|
Abeba™
31001/41 |
41 |
946.10 SEK
1 pair |
Estimated Shipment: 18-04-2024
Log in to see stock. |
|||||
15455734
|
Abeba™
31001/42 |
42 |
946.10 SEK
1 pair |
Estimated Shipment: 18-04-2024
Log in to see stock. |
|||||
15465734
|
Abeba™
31001/43 |
43 |
946.10 SEK
1 pair |
Estimated Shipment: 18-04-2024
Log in to see stock. |
|||||
15475734
|
Abeba™
31001/44 |
44 |
946.10 SEK
1 pair |
Estimated Shipment: 18-04-2024
Log in to see stock. |
|||||
15425744
|
Abeba™
31001/45 |
45 |
946.10 SEK
1 pair |
Estimated Shipment: 18-04-2024
Log in to see stock. |
|||||
15435744
|
Abeba™
31001/46 |
46 |
946.10 SEK
1 pair |
Estimated Shipment: 18-04-2024
Log in to see stock. |
|||||
Description
- CE EN ISO 20345:2011 SB A, E, FO SRC
- Microfiber
- White
- Steel toe cap
- Slip resistance PU outsole sole
- ESD dissipative according to DIN EN 61340
- Heel strap non-foldable, adjustable
- Microfiber lining and heel patch
- Adjustable instep area
- Buckle fastening
- Comfortable shape
Specifications
CE EN ISO 20345:2011 SB A, E, FO SRC | |
Microfiber | |
Slip-resistant | |
31001 | |
Animal fats, oil and fuel |
White | |
Microfiber | |
Polyurethane | |
Steel |