missing translation for 'onlineSavingsMsg'
Learn More

Tenak™ Classic Rack for Chest Freezers, 75 mm Box Height

Classic Rack is suitable for use with all chest freezer models.

Brand:  Tenak™ TE21462

Additional Details : Weight : 1.13400kg

 View more versions of this product

Product Code. 15280494

  • 925.00 SEK / Each
Estimated Shipment: 21-05-2024
to see stock.
Add to basket

Description

Description

  • Comes with a locking rod to secure the cryoboxes
  • Measure the inside height of you freezer when choosing the right rack
  • Constructed of stainless steel
Specifications

Specifications

Locking rod
Stainless Steel
140 x 140 x 486 mm
Boxes (6)
Chest Frseezers
75 mm (Box)
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers