Learn More
Tenak™ Classic Rack for Chest Freezers, 75 mm Box Height, with Boxes
Classic Rack is suitable for use with all chest freezer models.
890.00 SEK - 1550.00 SEK
Specifications
Includes | White cardboard boxes, 9 x 9 cell dividers |
---|---|
Material | Stainless Steel |
For Use With (Equipment) | Chest Frseezers |
Height (Metric) | 75 mm (Box) |
Product Code | Brand | Holds | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Holds | Price | Quantity & Availability | |||||
15200494
|
Tenak™
TE21455B |
Boxes (2) |
890.00 SEK
Each |
Estimated Shipment: 15-07-2024
Log in to see stock. |
|||||
15210504
|
Tenak™
TE21464B |
Boxes (7) |
1355.00 SEK
Each |
Estimated Shipment: 12-07-2024
Log in to see stock. |
|||||
15220504
|
Tenak™
TE21466B |
Boxes (8) |
1440.00 SEK
Each |
Estimated Shipment: 12-07-2024
Log in to see stock. |
|||||
15230494
|
Tenak™
TE21457B |
Boxes (3) |
975.00 SEK
Each |
Estimated Shipment: 12-07-2024
Log in to see stock. |
|||||
15230504
|
Tenak™
TE21467B |
Boxes (9) |
1550.00 SEK
Each |
Estimated Shipment: 12-07-2024
Log in to see stock. |
|||||
15250494
|
Tenak™
TE21458B |
Boxes (4) |
1070.00 SEK
Each |
Estimated Shipment: 12-07-2024
Log in to see stock. |
|||||
15270494
|
Tenak™
TE21460B |
Boxes (5) |
1150.00 SEK
Each |
Estimated Shipment: 12-07-2024
Log in to see stock. |
|||||
15290494
|
Tenak™
TE21462B |
Boxes (6) |
1245.00 SEK
Each |
Estimated Shipment: 12-07-2024
Log in to see stock. |
|||||
Description
- Comes with a locking rod to secure the cryoboxes
- Measure the inside height of you freezer when choosing the right rack
- Constructed of stainless steel
- Includes standard white cardboard boxes with 9 x 9 cell dividers
Specifications
White cardboard boxes, 9 x 9 cell dividers | |
Stainless Steel | |
Chest Frseezers | |
75 mm (Box) |