Learn More
Tenak™ Classic Rack for Chest Freezers, 85 mm Box Height
Classic Rack is suitable for use with all chest freezer models.
Brand: Tenak™ TE21465
Additional Details : Weight : 1.48000kg
Description
- Comes with a locking rod to secure the cryoboxes
- Measure the inside height of you freezer when choosing the right rack
- Constructed of stainless steel
Specifications
Locking rod | |
Stainless Steel | |
140 x 140 x 607 mm |
Boxes (7) | |
Chest Frseezers | |
85 mm (Box) |