Läs mer
Clavulanic Acid 2-Amino-2,4,4-trimethylpentane Salt, TRC
Produktkod. 30354205
Change view
Kvantitet:
5 mg
Produktkod. Kvantitet
30354205 5 mg
1 options
Produktkod. 30354205 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC5637555MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Clavulanic Acid 2-Amino-2,4,4-trimethylpentane Salt
CAS 66069-32-7
Molekylformel C8H9NO5 . C8H19N
Kvantitet 5 mg
Synonym [2R-(2alpha,3Z,5alpha)]-3-(2-Hydroxyethylidene)-7-oxo-4-oxa-1-azabicyclo[3.2.0]heptane-2-carboxylic Acid, compd. with 2,4,4-Trimethyl-2-pentanamine (1:1), 2,4,4-Trimethyl-2-pentanamine (2R,3Z,5R)-3-(2-Hydroxyethylidene)-7-oxo-4-oxa-1-azabicyclo[3.2.0]heptane-2-carboxylate, 2,4,4-Trimethyl-2-pentanamine [2R-(2alpha,3Z,5alpha)]-3-(2-Hydroxyethylidene)-7-oxo-4-oxa-1-azabicyclo[3.2.0]heptane-2-carboxylate
InChI formel InChI=1S/C9H11NO5/c1-14-9(13)8-5(2-3-11)15-7-4-6(12)10(7)8/h2,7-8,11H,3-4H2,1H3/b5-2-/t7-,8-/m1/s1
LEDER CC(C)(C)CC(C)(C)N.OC\C=C\1/O[C@@H]2CC(=O)N2[C@H]1C(=O)O
IUPAC-namn methyl (2R,3Z,5R)-3-(2-hydroxyethylidene)-7-oxo-4-oxa-1-azabicyclo[3.2.0]heptane-2-carboxylate
Molekylvikt (g/mol) 213.19
Formel vikt 213.0637