Läs mer
Clavulanic Acid Methyl Ester, TRC
Produktkod. 30360178
Change view
Kvantitet:
5 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30360178 50 mg
30363851 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30360178 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC56377050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 57943-82-5
Molekylformel C9 H11 N O5
Molekylvikt (g/mol) 213.19
Synonym 4-Oxa-1-azabicyclo[3.2.0]heptane-2-carboxylic acid, 3-(2-hydroxyethylidene)-7-oxo-, methyl ester, (2R,3Z,5R)-, 4-Oxa-1-azabicyclo[3.2.0]heptane-2-carboxylic acid, 3-(2-hydroxyethylidene)-7-oxo-, methyl ester, [2R-(2α,3Z,5α)]- (9CI), Methyl clavulanate
IUPAC-namn methyl (2R,3Z,5R)-3-(2-hydroxyethylidene)-7-oxo-4-oxa-1-azabicyclo[3.2.0]heptane-2-carboxylate
LEDER COC(=O)[C@@H]1N2[C@@H](CC2=O)O/C/1=C\CO
Specifikationer
Kemiskt namn eller material Clavulanic acid methyl ester
Kvantitet 50 mg
InChI formel InChI=1S/C9H11NO5/c1-14-9(13)8-5(2-3-11)15-7-4-6(12)10(7)8/h2,7-8,11H,3-4H2,1H3/b5-2-/t7-,8-/m1/s1
Formel vikt 213.0637