Beskrivning
As an integral part of the 3M™ Clean-Trace™ Hygiene Management System, the Clean-Trace Water Test uses adenosine tri-phosphate (ATP) to monitor the outcomes of manual cleaning of flexible endoscopes prior to sterilization or high-level disinfection.
- Simple to easy
- Powerful addition to your hygiene program
- Results measured in the 3M™ Clean-Trace™ NG Luminometer and provided in Relative Light Units (RLU) to allow rapid decisions on cleanliness status
Specifikationer
Specifikationer
| För användning med (applikation) | Water Testing for Surgical Instruments and Environmental Surfaces |
| Antal per förpackning | 100 Pk. |
| Produkttyp | Hygiene Management System |