Learn More
Phenomenex™ Cleanert™ Screw Caps with Septa, 9 mm, Pre-Slit
Product Code. 31745020 Shop All Phenomenex Products
Change view
Click to view available options
Color:
Blue
Unit Size:
Pack of 100
Product Code. Color Unit Size
31745020 Blue Pack of 100
1 options
Product Code. 31745020 Supplier Phenomenex™ Supplier No. AV22000
Beräknad leverans: 04-12-2026
Accessories for use with chromatography vials and in various chromatography applications.
  • Brand: Cleanert
  • Product: Accessory
TRUSTED_SUSTAINABILITY
Specifications
Color Blue
Material Polypropylene
Pre-slit Yes
Sterility Sterile
Septum PTFE/Silicone
Closure Size 9 mm
Marking Spot No
Autoclavable Autoclavable
Product Type Cap
Septum Color White/Red