Learn More
Fisherbrand™ Clear Straight Sided Glass Jars without Caps
GREENER_CHOICE
Catalog Number p-7144778
Click to view available options
Capacity (English):
1 oz.
2 oz.
4 oz.
8 oz.
16 oz.
32 oz.
Capacity (Metric):
30 mL
60 mL
120 mL
240 mL
480 mL
Clear, USP Type III soda-lime glass wide mouth design is great for large solid samples and allow for complete removal of contents. Ideal for soil sampling and environmental applications
  • Straight sided walls allow for the complete removal of contents
  • Available in a wide variety of sizes, from 1 to 32 oz.
Closure Size 43-400 mm
Diameter (Metric) 43 mm
Height (Metric) 43 mm
Product Type Straight Sided Jar
Capacity (Metric) 30 mL
Capacity (English) 1 oz.
Color Clear
Diameter (Metric) Outer Neck 39 mm
Diameter (Metric) Inner Neck 28 mm
Material Glass
Quantity 384
Neck Finish 43-400
Clarity Clear
Closure Included No
Mouth Wide
Shape Round