missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CLEC16A Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 26257-1-AP-20UL
Additional Details : Weight : 0.00010kg
Description
This gene encodes a member of the C-type lectin domain containing family. Single nucleotide polymorphisms in introns of this gene have been associated with diabetes mellitus, multiple sclerosis and rheumatoid arthritis. Multiple transcript variants encoding different isoforms have been found for this gene.Specifications
CLEC16A | |
Polyclonal | |
Unconjugated | |
CLEC16A | |
CLEC16A, Gop 1, KIAA0350, Protein CLEC16A | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
23274, 74374 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.35 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q2KHT3, Q80U30 | |
Clec16a | |
CLEC16A Fusion Protein Ag24268 | |
20 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |