Läs mer
Clenbuterol Hydrochloride, TRC
Produktkod. 30352649
Change view
Kvantitet:
100 mg
Produktkod. Kvantitet
30352649 100 mg
1 options
Produktkod. 30352649 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC569998100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Clenbuterol hydrochloride
CAS 21898-19-1
Anteckningar om renhetsgrad HPLC
Molekylformel C12 H18 Cl2 N2 O . Cl H
Kvantitet 100 mg
Synonym Benzenemethanol, 4-amino-3,5-dichloro-α-[[(1,1-dimethylethyl)amino]methyl]-, monohydrochloride (9CI), Benzyl alcohol, 4-amino-α-[(tert-butylamino)methyl]-3,5-dichloro-, monohydrochloride (8CI), 1-(4-Amino-3,5-dichlorophenyl)-2-(tert-butylamino)ethanol hydrochloride, 4-Amino-α-(tert-butylaminomethyl)-3,5-dichlorobenzyl alcohol monohydrochloride, Clenasma, Clenbuterol hydrochloride, NAB 365, NAB 365Cl, Spiropent, Ventipulmin
InChI formel InChI=1S/C12H18Cl2N2O.ClH/c1-12(2,3)16-6-10(17)7-4-8(13)11(15)9(14)5-7;/h4-5,10,16-17H,6,15H2,1-3H3;1H
LEDER Cl.CC(C)(C)NCC(O)c1cc(Cl)c(N)c(Cl)c1
IUPAC-namn 1-(4-amino-3,5-dichlorophenyl)-2-(tert-butylamino)ethanol;hydrochloride
Molekylvikt (g/mol) 313.65
Formel vikt 312.0563