Läs mer
Clenisopenterol Hydrochloride, TRC
Produktkod. 30138007
Change view
Kvantitet:
5 mg
10 mg
25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30138007 25 mg
30005744 5 mg
30004688 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30138007 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC57270025MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1435935-00-4
Molekylformel C13 H20 Cl2 N2 O . Cl H
Molekylvikt (g/mol) 327.68
Synonym Clenisopenterol hydrochloride, Benzenemethanol, 4-amino-3,5-dichloro-α-[[(3-methylbutyl)amino]methyl]-, hydrochloride (1:1)
IUPAC-namn 1-(4-amino-3,5-dichlorophenyl)-2-(3-methylbutylamino)ethanol;hydrochloride
LEDER Cl.CC(C)CCNCC(O)c1cc(Cl)c(N)c(Cl)c1
Specifikationer
Kemiskt namn eller material Clenisopenterol Hydrochloride
Kvantitet 25 mg
Anteckningar om renhetsgrad HPLC
InChI formel InChI=1S/C13H20Cl2N2O.ClH/c1-8(2)3-4-17-7-12(18)9-5-10(14)13(16)11(15)6-9;/h5-6,8,12,17-18H,3-4,7,16H2,1-2H3;1H
Formel vikt 326.0719
Procent renhet >95%