Läs mer
Clethodim Sulfoxide, TRC
Produktkod. 17990665
Change view
Kvantitet:
5 mg
10 mg
25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
17990665 25 mg
17923785 5 mg
17983185 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 17990665 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC57327025MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 111031-14-2
Molekylformel C17 H26 Cl N O4 S
Molekylvikt (g/mol) 375.91
Synonym 2-Cyclohexen-1-one, 2-[1-[[(3-chloro-2-propenyl)oxy]imino]propyl]-5-[2-(ethylsulfinyl)propyl]-3-hydroxy- (9CI), 2-[1-[[(3-Chloro-2-propen-1-yl)oxy]imino]propyl]-5-[2-(ethylsulfinyl)propyl]-3-hydroxy-2-cyclohexen-1-one, Clethodim sulfoxide
IUPAC-namn 2-[(E)-N-[(E)-3-chloroprop-2-enoxy]-C-ethylcarbonimidoyl]-5-(2-ethylsulfinylpropyl)-3-hydroxycyclohex-2-en-1-one
LEDER CC\C(=N/OC\C=C\Cl)\C1=C(O)CC(CC(C)S(=O)CC)CC1=O
Specifikationer
Kemiskt namn eller material Clethodim Sulfoxide
Kvantitet 25 mg
InChI formel InChI=1S/C17H26ClNO4S/c1-4-14(19-23-8-6-7-18)17-15(20)10-13(11-16(17)21)9-12(3)24(22)5-2/h6-7,12-13,20H,4-5,8-11H2,1-3H3/b7-6+,19-14+
Formel vikt 375.1271