Läs mer
Cletoquine Hydrochloride, TRC
Produktkod. 30343519
Change view
Kvantitet:
25 mg
Produktkod. Kvantitet
30343519 25 mg
1 options
Produktkod. 30343519 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC11777025MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Cletoquine Hydrochloride
CAS 159358-29-9
Molekylformel C16 H22 Cl N3 O • x HCl
Kvantitet 25 mg
Synonym 2-[[4-[(7-Chloro-4-quinolinyl)amino]pentyl]amino]-ethanol Hydrochloride,2-((4-((7-Chloroquinolin-4-yl)amino)pentyl)amino)ethanol Hydrochloride,Hydroxychloroquine Sulfate EP Impurity C
InChI formel InChI=1S/C16H22ClN3O.ClH/c1-12(3-2-7-18-9-10-21)20-15-6-8-19-16-11-13(17)4-5-14(15)16;/h4-6,8,11-12,18,21H,2-3,7,9-10H2,1H3,(H,19,20);1H
LEDER ClC1=CC2=NC=CC(NC(C)CCCNCCO)=C2C=C1.[xHCl]
IUPAC-namn 2-((4-((7-chloroquinolin-4-yl)amino)pentyl)amino)ethan-1-ol hydrochloride
Molekylvikt (g/mol) 307.82
Formel vikt 307.15