Läs mer
Tocris Bioscience™ Click N-Acetylmuramic acid - alkyne
Beskrivning
Derivative of N-acetylmuramic acid (NAM) component of bacterial peptidoglycans. Incorporated into bacterial peptidoglycans during biosynthesis. Suitable for fluorescent labeling of peptidoglycans when click-conjugated to a fluorescent dye.
Specifikationer
Specifikationer
| Kemiskt namn eller material | (2R)-2-(((3R,4R,5S,6R)-2,5-dihydroxy-6-(hydroxymethyl)-3-(pent-4-ynamido)tetrahydro-2H-pyran-4-yl)oxy)propanoic acid |
| CAS | 2245794-65-2 |
| Kvantitet | 2 mg |
| Mål | Glycobiology Probes |
| Molekylformel | C14H21NO8 |
| Renhet | 95% |