missing translation for 'onlineSavingsMsg'
Learn More

Nickel Electro™ Clifton™ Digital Thermostirrer with Stainless Steel Tank and Shelf

Get optimum water circulation and temperature stability with the digital thermostirrer. Designed for immersion of flasks, bottles, racks in a highly stable temperature environment, this Clifton™ Digital Thermostirrer with Stainless Steel Tank and Shelf includes “anti-bacterial technology” and other unique features as demanded by many clinical and laboratory environments.

POA

Specifications

Temperature Range 5°C to 99°C
Display LED
Voltage 230 V
Frequency 50/60 Hz
View More Specs

Description

Description

  • Designed for immersion of flasks, bottles, racks in an accurate temperature environment
  • Features digital PID temperature control
  • Immersed components are corrosion resistant
  • Thermostirrers are robustly constructed from stainless steel for durability
  • Easy to use controls that wipe clean and have an anti-bacterial finish
  • Thermostirrer controls and painted surfaces feature “anti-bacterial technology” 
  • When low liquid level is detected, a “FiLL” message automatically appears and heating is suspended
  • Every unit undergoes a factory 2 point calibration procedure 
Specifications

Specifications

5°C to 99°C
LED
230 V
50/60 Hz
Videos
SDS
Documents

Documents

Product Certifications