Learn More
Nickel Electro™ Clifton™ Stainless Steel Tank
Stainless Steel Tank providing an external vessel of circulating temperature controlled water.
Brand: Nickel Electro™ NE4-22U
Additional Details : Weight : 0.80000kg
Description
- Double-walled construction
- External surfaces have an anti-bacterial paint finish
- Inner tank is a bright stainless steel.
- While in use, submerge Inlet and outlet hoses under water from NE6-FH Clifton Flow Heater.
Specifications
22 L | |
Stainless Steel Tank | |
Stainless Steel, Antibacterial Paint Finish |
Clifton Thermostirrer | |
Laboratory Use |