Learn More
Nickel Electro™ Clifton™ Stainless Steel Tank
Stainless Steel Tank providing an external vessel of circulating temperature controlled water.
Brand:  Nickel Electro™ NE4-14U
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 11726587
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Description
Description
  • Double-walled construction
  • External surfaces have an anti-bacterial paint finish
  • Inner tank is a bright stainless steel.
  • While in use, submerge Inlet and outlet hoses under water from NE6-FH Clifton Flow Heater.
Specifications
Specifications
14 L
Stainless Steel Tank
Stainless Steel, Antibacterial Paint Finish
Clifton Thermostirrers
Laboratory Use