Läs mer
MMM Medcenter™ Climacell Climatic Cabinet, 222 L
Kapacitet (metrisk):
222 l
Förpackningsstorlek:
Varje
Beskrivning
- Forced ventilation of air and cooling with controlled humidity
- All environmental conditions can be adapted with its precise control of humidity and moisture
- Temperature range: 0 to 99.9°C
- Microprocessor control
- Chipcard system for storing of the programmes
- Interface RS 232
- Audible alarm
- Integral timer function up to max. 16 years
- Digital protective thermostat class 3
- Programming of ramps, cycles
- Real time
Specifikationer
Specifikationer
| Kapacitet (metrisk) | 222 l |
| Mått (D x B x H) Inredning | 520 x 540 x 760 mm |
| Mått (L x B x H) | 790 x 760 x 1230 mm |