Learn More
Climbazole, TRC
Product Code. 30368066
Change view
Click to view available options
Quantity:
10 mg
50 mg
25 g
Unit Size:
10mg
25g
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30368066 50 mg 50mg
30361934 25 g 25g
30365387 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30368066 Supplier Toronto Research Chemicals Supplier No. TRCC57850050MG
 Restricted Item
Beräknad leverans: 09-12-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 38083-17-9
Molecular Formula C15 H17 Cl N2 O2
Molecular Weight (g/mol) 292.76
Synonym 1-(4-Chlorophenoxy)-1-(1H-imidazol-1-yl)-3,3-dimethyl-2-butanone, 1-(4-Chlorophenoxy)-1-(imidazol-1-yl)-3,3-dimethylbutan-2-one, 1-[(4-Chlorophenoxy)(tert-butylcarbonyl)methyl]imidazole, BAY-e 6975, Baypival, Climbazole, Crinipan AD, Diazolone
IUPAC Name 1-(4-chlorophenoxy)-1-imidazol-1-yl-3,3-dimethylbutan-2-one
SMILES CC(C)(C)C(=O)C(Oc1ccc(Cl)cc1)n2ccnc2
Specifications
Quantity 50 mg
InChI Formula InChI=1S/C15H17ClN2O2/c1-15(2,3)13(19)14(18-9-8-17-10-18)20-12-6-4-11(16)5-7-12/h4-10,14H,1-3H3
Formula Weight 292.0979
Chemical Name or Material Climbazole