Läs mer
Clindamycin 2-Phosphate, TRC
Specifikationer
Specifikationer
| Kemiskt namn eller material | Clindamycin phosphate |
| CAS | 24729-96-2 |
| Molekylformel | C18 H34 Cl N2 O8 P S |
| Kvantitet | 1 g |
| Synonym | Clindamycin Phosphate, L-threo-α-D-galacto-Octopyranoside, methyl 7-chloro-6,7,8-trideoxy-6-[[[(2S,4R)-1-methyl-4-propyl-2-pyrrolidinyl]carbonyl]amino]-1-thio-, 2-(dihydrogen phosphate), L-threo-D-galacto-Octopyranoside, methyl 7-chloro-6,7,8-trideoxy-6-(1-methyl-4-propyl-L-2-pyrrolidinecarboxamido)-1-thio-, 2-(dihydrogen phosphate), trans- α-, L-threo-α-D-galacto-Octopyranoside, methyl 7-chloro-6,7,8-trideoxy-6-[[(1-methyl-4-propyl-2-pyrrolidinyl)carbonyl]amino]-1-thio-, 2-(dihydrogen phosphate), (2S-trans)-, 7(S)-Chloro-7-deoxylincomycin 2-phosphate, Cleocin T, Cleocin phosphate, Clinadac, Clindac, Clindagel, Clindamycin 2-phosphate, Clindesse, Clindets, Dalacin P, Dalacin T, Methyl 7-chloro-6,7,8-trideoxy-6-trans-(1-methyl-4-propyl-L-2-pyrrolidinecarboxamido)-1-thio-L-threo-α-D-galacto-octopyranoside 2-(dihydrogen phosphate), NSC 618653, U 28508 |
| InChI formel | InChI=1S/C18H34ClN2O8PS/c1-5-6-10-7-11(21(3)8-10)17(24)20-12(9(2)19)15-13(22)14(23)16(18(28-15)31-4)29-30(25,26)27/h9-16,18,22-23H,5-8H2,1-4H3,(H,20,24)(H2,25,26,27)/t9-,10+,11-,12+,13+,14-,15+,16+,18+/m0/s1 |
| LEDER | CCC[C@@H]1C[C@H](N(C)C1)C(=O)N[C@H]([C@H](C)Cl)[C@H]2O[C@H](SC)[C@H](OP(=O)(O)O)[C@@H](O)[C@H]2O |
| IUPAC-namn | [(2R,3R,4S,5R,6R)-6-[(1S,2S)-2-chloro-1-[[(2S,4R)-1-methyl-4-propylpyrrolidine-2-carbonyl]amino]propyl]-4,5-dihydroxy-2-methylsulfanyloxan-3-yl] dihydrogen phosphate |
| Molekylvikt (g/mol) | 504.96 |
| Formel vikt | 504.1462 |