Läs mer
Clindamycin 3-Palmitate Hydrochloride, TRC
Produktkod. 30357858
Change view
Kvantitet:
25 mg
Produktkod. Kvantitet
30357858 25 mg
1 options
Produktkod. 30357858 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC58006025MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Clindamycin 3-Palmitate Hydrochloride
CAS 30747-19-4
Molekylformel C34H63ClN2O6S •HCl
Kvantitet 25 mg
Synonym (2S-trans)-Methyl 7-Chloro-6,7,8-trideoxy-6-[[(1-methyl-4-propyl-2-pyrrolidinyl)carbonyl]amino]-1-thio-L-threo-α-D-galacto-octopyranoside 3-Hexadecanoate Hydrochloride
InChI formel InChI=1S/C34H63ClN2O6S.ClH/c1-6-8-9-10-11-12-13-14-15-16-17-18-19-21-27(38)42-32-29(39)31(43-34(44-5)30(32)40)28(24(3)35)36-33(41)26-22-25(20-7-2)23-37(26)4;/h24-26,28-32,34,39-40H,6-23H2,1-5H3,(H,36,41);1H/t24-,25+,26+,28+,29-,30+,31+,32-,34+;/m0./s1
LEDER O[C@H]1[C@@H]([C@H](NC(C2C[C@@H](CCC)CN2C)=O)[C@@H](Cl)C)O[C@H](SC)[C@H](O)[C@H]1OC(CCCCCCCCCCCCCCC)=O.Cl
IUPAC-namn (2R,3S,4S,5R,6R)-2-((1S,2S)-2-chloro-1-((4R)-1-methyl-4-propylpyrrolidine-2-carboxamido)propyl)-3,5-dihydroxy-6-(methylthio)tetrahydro-2H-pyran-4-yl palmitate hydrogen chloride
Molekylvikt (g/mol) 699.85
Formel vikt 698.4