Läs mer
Clindamycin Palmitate Sulfoxide Hydrochloride (>90%), TRC
Produktkod. 30351746
Change view
Kvantitet:
5 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30351746 5 mg
30364333 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30351746 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC5800255MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 2126928-92-3
Molekylformel C34 H63 Cl N2 O7 S . Cl H
Molekylvikt (g/mol) 715.85
Synonym Clindamycin Palmitate Sulfoxide Hydrochloride, 7-Chloro-1,6,7,8-tetradeoxy-6-[[[(2S,4R)-1-methyl-4-propyl-2-pyrrolidinyl]carbonyl]amino]-1-(methylsulfinyl)-L-threo-α-D-galacto-octopyranose 2-hexadecanoate hydrochloride
IUPAC-namn [(2R,3R,4S,5R,6R)-6-[(1S,2S)-2-chloro-1-[[(2S,4R)-1-methyl-4-propylpyrrolidine-2-carbonyl]amino]propyl]-4,5-dihydroxy-2-methylsulfinyloxan-3-yl] hexadecanoate;hydrochloride
LEDER Cl.CCCCCCCCCCCCCCCC(=O)O[C@@H]1[C@@H](O)[C@@H](O)[C@H](O[C@@H]1S(=O)C)[C@H](NC(=O)[C@@H]2C[C@@H](CCC)CN2C)[C@H](C)Cl
Specifikationer
Kemiskt namn eller material Clindamycin Palmitate Sulfoxide Hydrochloride
Kvantitet 5 mg
InChI formel InChI=1S/C34H63ClN2O7S.ClH/c1-6-8-9-10-11-12-13-14-15-16-17-18-19-21-27(38)43-32-30(40)29(39)31(44-34(32)45(5)42)28(24(3)35)36-33(41)26-22-25(20-7-2)23-37(26)4;/h24-26,28-32,34,39-40H,6-23H2,1-5H3,(H,36,41);1H/t24-,25+,26-,28+,29+,30-,31+,32+,34+,45?;/m0./s1
Formel vikt 714.3811