Läs mer
Clioquinol, LoGiCal
Produktkod. 30461445
Change view
Kvantitet:
10 mg
Produktkod. Kvantitet
30461445 10 mg
1 options
Produktkod. 30461445 Leverantör LoGiCal Leverantörsnummer LGCFOR0312.00
LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Clioquinol
CAS 130-26-7
Analyt- eller komponentnamn Clioquinol
Frakt skick Room Temperature
Molekylformel C9 H5 Cl I N O
Certifikat och efterlevnad ISO 9001
Synonym Clioquinol, 5-Chloro-7-iodo-8-hydroxyquinoline, 5-Chloro-7-iodo-8-oxyquinoline, 5-Chloro-7-iodo-8-quinolinol, 5-Chloro-8-hydroxy-7-iodoquinoline, 7-Iodo-5-chloro-8-hydroxyquinoline, 7-Iodo-5-chloroxine, 8-Hydroxy-7-iodo-5-chloroquinoline, Alchloquin, Amebil, Amoenol, Bactol, Barquinol, Budoform, Chinoform, Chloroiodoquin, Chloroiodoquine, Chlorojodochin, Cifoform, Cliquinol, Cort-Quin, Dioquinol, Dizenterol, Eczecidin, Emaform, Entero-Vioform, Enteroquinol, Enteroseptol, Enterozol, Entrokin, Hi-Eneterol, Hi-Enterol, Iodenterol, Iodochlorhydroxyquin, Iodochlorhydroxyquinol, Iodochloroquine, Iodochloroxine, Iodochloroxyquinoline, Iodoenterol, Iodoxyquinoline, Lekosept, NSC 3531, NSC 74938, Nioform, Quin-O-Creme, Quinambicide, Quinidochlor, Quiniodochlor, Quinoform, Quinoform (antiseptic), Rheaform, Rheaform Boluses, Rometin, Vioform, Vioformio
InChI formel InChI=1S/C9H5ClINO/c10-6-4-7(11)9(13)8-5(6)2-1-3-12-8/h1-4,13H
Lösningstyp Neat
LEDER Oc1c(I)cc(Cl)c2cccnc12
IUPAC-namn 5-chloro-7-iodoquinolin-8-ol
Industry Type Clinical Testing
Molekylvikt (g/mol) 305.5
Kvantitet 10 mg
Formel vikt 304.9104 g/mol