Läs mer
Clioquinol, Mikromol™
Beskrivning
Clioquinol
Specifikationer
Specifikationer
| Kemiskt namn eller material | Clioquinol |
| CAS | 130-26-7 |
| Analyt- eller komponentnamn | Clioquinol |
| Frakt skick | Room Temperature |
| Molekylformel | C9 H5 Cl I N O |
| Certifikat och efterlevnad | ISO 17034 |
| Synonym | Clioquinol, 5-Chloro-7-iodo-8-hydroxyquinoline, 5-Chloro-7-iodo-8-oxyquinoline, 5-Chloro-7-iodo-8-quinolinol, 5-Chloro-8-hydroxy-7-iodoquinoline, 7-Iodo-5-chloro-8-hydroxyquinoline, 7-Iodo-5-chloroxine, 8-Hydroxy-7-iodo-5-chloroquinoline, Alchloquin, Amebil, Amoenol, Bactol, Barquinol, Budoform, Chinoform, Chloroiodoquin, Chloroiodoquine, Chlorojodochin, Cifoform, Cliquinol, Cort-Quin, Dioquinol, Dizenterol, Eczecidin, Emaform, Entero-Vioform, Enteroquinol, Enteroseptol, Enterozol, Entrokin, Hi-Eneterol, Hi-Enterol, Iodenterol, Iodochlorhydroxyquin, Iodochlorhydroxyquinol, Iodochloroquine, Iodochloroxine, Iodochloroxyquinoline, Iodoenterol, Iodoxyquinoline, Lekosept, NSC 3531, NSC 74938, Nioform, Quin-O-Creme, Quinambicide, Quinidochlor, Quiniodochlor, Quinoform, Quinoform (antiseptic), Rheaform, Rheaform Boluses, Rometin, Vioform, Vioformio |
| InChI formel | InChI=1S/C9H5ClINO/c10-6-4-7(11)9(13)8-5(6)2-1-3-12-8/h1-4,13H |
| LEDER | Oc1c(I)cc(Cl)c2cccnc12 |
| IUPAC-namn | 5-chloro-7-iodoquinolin-8-ol |
| Visa mer |