Learn More
Fisherbrand™ Clips
Clips are ideal for easy loading and removal of tubes.
37.85 SEK - 91.50 SEK
Product Code | Brand | For Use With (Equipment) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | For Use With (Equipment) | Price | Quantity & Availability | |||||
12318458
|
Fisherbrand™
|
For 15 to 17 mm Tubes |
91.50 SEK
Each |
Estimated Shipment: 30-07-2024
Log in to see stock. |
|||||
12328458
|
Fisherbrand™
|
For 27 to 30 mm Tubes |
91.50 SEK
Each |
Estimated Shipment: 30-07-2024
Log in to see stock. |
|||||
12308458
|
Fisherbrand™
|
For 8 to 11 mm Tubes |
37.85 SEK
Each |
Estimated Shipment: 30-07-2024
Log in to see stock. |
|||||
Description
- Supplied in singles
- The range of accessories can also be autoclaved and allow for easy loading and tube removal
Specifications
Clip |