missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CLIP1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 23839-1-AP-20UL
Additional Details : Weight : 0.00010kg
Description
This gene encodes a protein which specifically interacts with translin, a DNA-binding protein that binds consensus sequences at breakpoint junctions of chromosomal translocations. The encoded protein contains bipartite nuclear targeting sequences that may provide nuclear transport for translin, which lacks any nuclear targeting motifs.Specifications
CLIP1 | |
Polyclonal | |
Unconjugated | |
CLIP1 | |
CLIP, CLIP 170, CLIP1, CLIP170, CYLN1, Cytoplasmic linker protein 1, Restin, RSN | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
56430, 6249, 65201 | |
-20°C | |
Liquid |
Immunocytochemistry, Western Blot, Immunofluorescence, Immunoprecipitation | |
0.51 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P30622, Q922J3, Q9JK25 | |
CLIP1 | |
CLIP1 Fusion Protein Ag20811 | |
20 μL | |
Primary | |
Mouse, Rat, Human | |
Antibody | |
IgG |