missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CLIP1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1656.73 SEK - 4721.67 SEK
Specifications
Antigen | CLIP1 |
---|---|
Concentration | 0.51 mg/mL |
Applications | Immunocytochemistry, Western Blot, Immunofluorescence, Immunoprecipitation |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16882465
|
Proteintech
23839-1-AP-20UL |
20 μL |
1656.73 SEK
20µL |
Estimated Shipment: 26-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16872465
|
Proteintech
23839-1-AP-150UL |
150 μL |
4721.67 SEK
150µL |
Estimated Shipment: 26-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This gene encodes a protein which specifically interacts with translin, a DNA-binding protein that binds consensus sequences at breakpoint junctions of chromosomal translocations. The encoded protein contains bipartite nuclear targeting sequences that may provide nuclear transport for translin, which lacks any nuclear targeting motifs.Specifications
CLIP1 | |
Immunocytochemistry, Western Blot, Immunofluorescence, Immunoprecipitation | |
Unconjugated | |
Rabbit | |
Mouse, Rat, Human | |
P30622, Q922J3, Q9JK25 | |
56430, 6249, 65201 | |
CLIP1 Fusion Protein Ag20811 | |
Primary | |
-20°C | |
CLIP1 |
0.51 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
CLIP, CLIP 170, CLIP1, CLIP170, CYLN1, Cytoplasmic linker protein 1, Restin, RSN | |
CLIP1 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |