missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CLIP4 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 17044-1-AP-150UL
Additional Details : Weight : 0.00010kg
Description
CLIP4 includes 4 isoforms of the human protein that are produced by alternative splicing.Specifications
CLIP4 | |
Polyclonal | |
Unconjugated | |
CLIP4 | |
CLIP4, Restin like protein 2, RSNL2 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
79745 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Western Blot | |
0.41 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q8N3C7 | |
CLIP4 | |
CLIP4 Fusion Protein Ag10590 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |