Learn More
Thermo Scientific™ ClipTip™ 384-Format Pipette Tips
Ensure a secure seal for every sample, with ClipTip™ 384-format Pipette Tips that work well together with E1-ClipTip™. A unique, innovative interlocking technology ensures a complete seal on every channel with minimal tip attachment and ejection force.
3426.00 SEK - 4285.00 SEK
Specifications
For Use With (Equipment) Thermo Scientific™ E1-ClipTip
Product Line ClipTip
Filtered Non-filtered
View More Specs
Products 8
Product Code Brand Description Color Code Price Quantity & Availability  
Product Code Brand Description Color Code Price Quantity & Availability  
15206293
View Documents
Thermo Scientific™
94410050
ClipTip 384 12.5, nonsterile Pink
3446.00 SEK
Pack of 3840
View alternative products
Save Up to 
»
Estimated Shipment
04-01-2021
Add to basket
15962011
View Documents
Thermo Scientific™
94410070
ClipTip 384 12.5 Ext, nonsterile Pink
3426.00 SEK
Pack of 3840
View alternative products
Save Up to 
»
Estimated Shipment
04-01-2021
Add to basket
15972011
View Documents
Thermo Scientific™
94410073
ClipTip 384 12.5 Ext, sterile Pink
4254.00 SEK
Pack of 3840
View alternative products
Save Up to 
»
Estimated Shipment
12-05-2021
Add to basket
15216293
View Documents
Thermo Scientific™
94410053
ClipTip 384 12.5, sterile Pink
4028.00 SEK
Pack of 3840
View alternative products
Save Up to 
»
Estimated Shipment
04-01-2021
Add to basket
15236293
View Documents
Thermo Scientific™
94410100
ClipTip 384 30, nonsterile Violet
3637.00 SEK
Pack of 3840
View alternative products
Save Up to 
»
Estimated Shipment
15-02-2021
Add to basket
15246293
View Documents
Thermo Scientific™
94410103
ClipTip 384 30, sterile Violet
4285.00 SEK
Pack of 3840
View alternative products
Save Up to 
»
In stock, ready to ship within 7 days: 6
Add to basket
15266293
View Documents
Thermo Scientific™
94410150
ClipTip 384 125, nonsterile Yellow
3446.00 SEK
Pack of 3840
View alternative products
Save Up to 
»
Estimated Shipment
04-01-2021
Add to basket
15276293
View Documents
Thermo Scientific™
94410153
ClipTip 384 125, sterile Yellow
4285.00 SEK
Pack of 3840
Visualizzare prodotti alternativi
missing translation for 'toSaveUpto' 
»
Estimated Shipment
17-03-2021
Aggiungi al carrello
Descrizione
Descrizione
Key Features:
  • ClipTip pipette tips feature 'clips' that lock tips in place and forms a complete seal
  • Each time ClipTip pipette tips are on, they are sealed and will not loosen regardless of application pressure
  • ClipTip tip is clipped securely on the F1-ClipTip pipette with a light touch by eliminating the frustration of banging tips on your pipette
Low retention
  • Every ClipTip pipette tip utilizes low retention technology to enable maximium sample recover
Proven quality
  • Certified free of RNase, DNase, DNA, ATP and endotoxin contamination
Environmentally responsible
  • ClipTip reload system uses recycled components and 30% less plastic
Versatile Tip Range
  • Choose from five sizes: 20, 50, 200, 300 and 1000μL
Immediate traceability
  • Each tip insert has ink-jetted lot and volume information; the convenient color key makes it easy to match the color of the F1-ClipTip pipette plunger cap with the corresponding ClipTip pipette tip rack insert
Note:
Availability may vary by country.
Specifica
Specifica
Thermo Scientific™ E1-ClipTip
ClipTip
Non-filtered
SDS