missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CLK1 Recombinant Human Protein
CLK1 is a dual specificity kinase that phosphorylates PTP-1B and members of the SR family of splicing factors.
Brand: Invitrogen™ PR5259A
Additional Details : Weight : 0.01000kg
Description
CLK1 is thought to be involved in neuronal differentiation and mRNA splicing.
Specifications
NP_004062 | |
50mM Tris, pH 7.5, 150mM NaCl, 0.05mM EDTA, 0.05% Triton X-100, 2mM DTT, 50% glycerol | |
100μg | |
RUO - research use only | |
CLK1 | |
Human | |
GST Tag | |
Frozen |
Enzyme and Protein Activity Assay, Kinase Assay, Protein Assays and Analysis, Protein Biology | |
70.1kDa | |
Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. | |
Catalytic | |
CLK Kinase Family | |
Recombinant | |
E. coli |