missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
Invitrogen™ CLN5 Polyclonal Antibody

Beskrivning
CLN5 Polyclonal Antibody detects endogenous levels of CLN5 protein.
This gene is one of eight which have been associated with neuronal ceroid lipofuscinoses (NCL). Also referred to as Batten disease, NCL comprises a class of autosomal recessive, neurodegenerative disorders affecting children. The genes responsible likely encode proteins involved in the degradation of post-translationally modified proteins in lysosomes. The primary defect in NCL disorders is thought to be associated with lysosomal storage function.[provided by RefSeq, Oct 2008].
Specifikationer
Specifikationer
| Antigen | CLN5 |
| Användningsområden | ELISA, Western Blot |
| Klassificering | Polyclonal |
| Koncentration | 1 mg/mL |
| Konjugera | Unconjugated |
| Formulering | PBS with 50% glycerol, 0.5% BSA and 0.02% sodium azide; pH 7.4 |
| Gen | CLN5 |
| Genaccessionsnr. | O75503, Q3UMW8 |
| Gene Alias | A730075N08Rik; ceroid-lipofuscinosis; Ceroid-lipofuscinosis neuronal protein 5; ceroid-lipofuscinosis neuronal protein 5 homolog; Ceroid-lipofuscinosis neuronal protein 5 homolog, secreted form; Ceroid-lipofuscinosis neuronal protein 5, secreted form; ceroid-lipofuscinosis, neuronal 5; CLN5; NCL; Protein CLN5 |
| Gensymboler | CLN5 |
| Visa mer |
Produkttitel
Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.
Hittar du en möjlighet till förbättring?