Läs mer
Tocris Bioscience™ Clobenpropit dihydrobromide
Highly potent H3 antagonist and H4 partial agonist
Brand: Tocris Bioscience™ 0752/10
Denna artikel kan inte returneras.
Se returpolicy
Beskrivning
An extremely potent histamine H3 antagonist/inverse agonist (pA2=9.93). Also displays partial agonist activity at H4 receptors; induces eosinophil shape change with an EC50 of 3 nM.Specifikationer
| N-(4-Chlorobenzyl)-S-[3-(4(5)-imidazolyl)propyl]isothiourea dihydrobromide | |
| >99% | |
| Soluble to 100 mM in water |
| 145231-35-2 | |
| 10 mg |
Produkttitel
For Research Use Only