Läs mer
Clocortolone Pivalate, TRC
Produktkod. 30355833
Change view
Kvantitet:
10 mg
Produktkod. Kvantitet
30355833 10 mg
1 options
Produktkod. 30355833 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC58381010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Clocortolone Pivalate
CAS 34097-16-0
Molekylformel C27 H36 Cl F O5
Kvantitet 10 mg
Synonym (6α,11β,16α)-9-Chloro-21-(2,2-dimethyl-1-oxopropoxy)-6-fluoro-11-hydroxy-16-methylpregna-1,4-diene-3,20-dione, Pregna-1,4-diene-3,20-dione, 9-chloro-6α-fluoro-11β,21-dihydroxy-16α-methyl-, 21-pivalate (8CI), 9-Chloro-6α-fluoro, 11β,21-dihydroxy-16α-methylpregna-1,4-diene-3,20-dione 21-pivalate, CL 68, Clocortolone pivalate, Clocortolone trimethylacetate, Cloderm, Purantix, SH 863
InChI formel InChI=1S/C27H36ClFO5/c1-14-9-16-17-11-19(29)18-10-15(30)7-8-26(18,6)27(17,28)21(32)12-25(16,5)22(14)20(31)13-34-23(33)24(2,3)4/h7-8,10,14,16-17,19,21-22,32H,9,11-13H2,1-6H3/t14-,16+,17+,19+,21+,22-,25+,26+,27+/m1/s1
LEDER C[C@@H]1C[C@H]2[C@@H]3C[C@H](F)C4=CC(=O)C=C[C@]4(C)[C@@]3(Cl)[C@@H](O)C[C@]2(C)[C@H]1C(=O)COC(=O)C(C)(C)C
IUPAC-namn [2-[(6S,8S,9R,10S,11S,13S,14S,16R,17S)-9-chloro-6-fluoro-11-hydroxy-10,13,16-trimethyl-3-oxo-7,8,11,12,14,15,16,17-octahydro-6H-cyclopenta[a]phenanthren-17-yl]-2-oxoethyl] 2,2-dimethylpropanoate
Molekylvikt (g/mol) 495.02
Formel vikt 494.2235